Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927370.5 | XP_001927405.1 | 1453 | 64 | ||
| GenBank mRNA | AK232053.1 | - | 5 - 6 | 890 | 64 | |
| AK347662.1 | - | 5 - 6 | 842 | 64 | ||
| AK389525.1 | - | 5 - 6 | 928 | 64 | ||
More Information
Set Membership: |
|

IGBP1
Genomic Map