Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021093899.1 | XP_020949558.1 | 543 | 75 | ||
XM_021093900.1 | XP_020949559.1 | 719 | 75 | |||
GenBank mRNA | AK232113.1 | - | 4 - 5 | 595 | 75 | |
AK232724.1 | - | 4 - 5 | 533 | 75 | ||
AK232782.1 | - | 4 - 5 | 519 | 75 | ||
AK234408.1 | - | 4 - 5 | 519 | 75 | ||
AK345889.1 | - | 4 - 5 | 515 | 75 | ||
AK346132.1 | - | 4 - 5 | 519 | 75 | ||
AK390218.1 | - | 4 - 5 | 607 | 75 | ||
AK397032.1 | - | 3 - 4 | 270 | 75 | ||
AK400664.1 | - | 4 - 5 | 512 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
