Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928553.5 | XP_001928588.2 | 571 | 62 | ||
XM_005671577.3 | XP_005671634.1 | 521 | 62 | |||
XM_005671579.3 | XP_005671636.2 | 631 | 62 | |||
XM_021073764.1 | XP_020929423.1 | 376 | 62 | |||
GenBank mRNA | AK232147.1 | - | 4 - 5 | 509 | 62 | |
AK234536.1 | - | 3 - 4 | 350 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
