Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232153.1 | - | 1 | 221 | 165 | |
| AK235209.1 | - | 7 | 1205 | 163 | ||
| AK235969.1 | - | 6 | 1311 | 163 | ||
| AK236273.1 | - | 7 | 2018 | 165 | ||
| AK238865.1 | - | 6 | 1229 | 165 | ||
| AK238990.1 | - | 7 | 1156 | 165 | ||
| AK239123.1 | - | 6 | 1252 | 165 | ||
| AK351956.1 | - | 6 | 1234 | 165 | ||
| AK395269.1 | - | 1 | 512 | 164 | ||
| AK395422.1 | - | 5 | 2028 | 165 | ||
More Information
Set Membership: |
|

FMO4
Genomic Map