Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098608.1 | XP_020954267.1 | 929 | 61 | ||
| XM_021098609.1 | XP_020954268.1 | 911 | 61 | |||
| XM_021098610.1 | XP_020954269.1 | 2367 | 61 | |||
| GenBank mRNA | AK389924.1 | - | 5 - 6 | 784 | 61 | |
| AK399907.1 | - | 5 - 6 | 827 | 61 | ||
More Information
Set Membership: |
|

PACSIN1
Genomic Map