Assay Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355200.4 | XP_003355248.1 | 259 | 61 | ||
| XM_003355201.3 | XP_003355249.1 | 183 | 61 | |||
| XM_005655396.3 | XP_005655453.1 | 194 | 61 | |||
| XM_005655397.3 | XP_005655454.1 | 264 | 61 | |||
| XM_013988092.2 | XP_013843546.1 | 305 | 61 | |||
| GenBank mRNA | AK389989.1 | - | 2 - 3 | 323 | 61 | |
More Information
Set Membership: |
|

MRPS21
Genomic Map