Assay Details
| Gene Symbol: MAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134032.5 | XP_003134080.1 | 708 | 56 | ||
| XM_005672533.3 | XP_005672590.1 | 947 | 56 | |||
| GenBank mRNA | AK390004.1 | - | 3 - 4 | 489 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005672533.3 transcripts.
Set Membership: |
|

MAP1B
Genomic Map