Assay Details
Gene Symbol: STUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357643.4 | XP_003357691.1 | 652 | 79 | ||
GenBank mRNA | AK390056.1 | - | 2 - 4 | 665 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003357643.4 | XP_003357691.1 | 652 | 79 | ||
GenBank mRNA | AK390056.1 | - | 2 - 4 | 665 | 79 |