Assay Details
Gene Symbol: KCNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087841.1 | XP_020943500.1 | 1835 | 64 | ||
GenBank mRNA | AK390074.1 | - | 1 | 1521 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
