Assay Details
Gene Symbol: ZNF25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005670992.3 | XP_005671049.1 | 690 | 100 | ||
XM_005670993.3 | XP_005671050.1 | 2047 | 100 | |||
XM_013983246.2 | XP_013838700.1 | 2070 | 100 | |||
GenBank mRNA | AK390160.1 | - | 2 - 3 | 305 | 100 | |
AK396647.1 | - | 2 - 3 | 231 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005670992.3, XM_005670993.3, XM_013983246.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
