Assay Details
| Gene Symbol: ZNF25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670992.3 | XP_005671049.1 | 815 | 92 | ||
| XM_005670993.3 | XP_005671050.1 | 2172 | 92 | |||
| XM_013983246.2 | XP_013838700.1 | 2195 | 92 | |||
| XM_021072377.1 | XP_020928036.1 | 881 | 92 | |||
| XM_021072378.1 | XP_020928037.1 | 2261 | 92 | |||
| GenBank mRNA | AK390160.1 | - | 3 - 4 | 430 | 92 | |
| AK396647.1 | - | 3 - 4 | 356 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021072377.1, XM_021072378.1 transcripts.
Set Membership: |
|

ZNF25
Genomic Map