Assay Details
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134361.4 | XP_003134409.2 | 265 | 83 | ||
| GenBank mRNA | AK390258.1 | - | 1 - 2 | 232 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SRXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134361.4 | XP_003134409.2 | 265 | 83 | ||
| GenBank mRNA | AK390258.1 | - | 1 - 2 | 232 | 83 | |
Set Membership: |
|