Assay Details
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674571.3 | XP_005674628.2 | 265 | 61 | ||
XM_013985225.2 | XP_013840679.1 | 265 | 61 | |||
XM_021077713.1 | XP_020933372.1 | 265 | 61 | |||
GenBank mRNA | AK390349.1 | - | 2 - 3 | 211 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
