Assay Details
Gene Symbol: BEND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003128942.5 | XP_003128990.3 | 1146 | 88 | ||
XM_013978637.2 | XP_013834091.2 | 1146 | 88 | |||
GenBank mRNA | AK390386.1 | - | 2 - 3 | 1003 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
