Assay Details
Gene Symbol: CCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003361792.4 | XP_003361840.1 | 321 | 80 | ||
GenBank mRNA | AK391153.1 | - | 1 - 2 | 38 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003361792.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
