Assay Details
Gene Symbol: WIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013990591.2 | XP_013846045.2 | 4226 | 105 | ||
XM_021074983.1 | XP_020930642.1 | 3868 | 105 | |||
XM_021074984.1 | XP_020930643.1 | 4237 | 105 | |||
XM_021074985.1 | XP_020930644.1 | 3979 | 105 | |||
GenBank mRNA | AK391232.1 | - | 1 | 914 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
