Assay Details
| Gene Symbol: LOC100624149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005657947.3 | XP_005658004.1 | 387 | 82 | ||
| XM_021080929.1 | XP_020936588.1 | 340 | 82 | |||
| GenBank mRNA | AK391373.1 | - | 3 - 4 | 287 | 82 | |
More Information
Set Membership: |
|

LOC100624149
Genomic Map