Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925635.5 | XP_001925670.2 | 1705 | 67 | ||
XM_005671487.3 | XP_005671544.2 | 1805 | 67 | |||
XM_005671488.3 | XP_005671545.2 | 1280 | 67 | |||
XM_021073402.1 | XP_020929061.1 | 1537 | 67 | |||
GenBank mRNA | AK391386.1 | - | 8 - 9 | 887 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
