Assay Details
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129877.5 | XP_003129925.1 | 975 | 103 | ||
GenBank mRNA | AK391414.1 | - | 4 - 5 | 383 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
