Assay Details
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096734.1 | XP_020952393.1 | 2415 | 59 | ||
XM_021096735.1 | XP_020952394.1 | 2477 | 59 | |||
XM_021096736.1 | XP_020952395.1 | 2432 | 59 | |||
GenBank mRNA | AK391547.1 | - | 3 - 4 | 417 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
