Assay Details
| Gene Symbol: DUSP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924537.5 | XP_001924572.2 | 996 | 87 | ||
| GenBank mRNA | AK391742.1 | - | 5 - 6 | 895 | 87 | |
More Information
Set Membership: |
|

DUSP12
Genomic Map