Assay Details
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129592.5 | XP_003129640.2 | 281 | 66 | ||
XM_005653676.3 | XP_005653733.1 | 275 | 66 | |||
XM_005653677.3 | XP_005653734.1 | 269 | 66 | |||
XM_005653678.3 | XP_005653735.1 | 1011 | 66 | |||
XM_005653680.3 | XP_005653737.1 | 413 | 66 | |||
XM_005653681.3 | XP_005653738.1 | 266 | 66 | |||
XM_005653682.3 | XP_005653739.1 | 282 | 66 | |||
XM_013999319.2 | XP_013854773.1 | 1015 | 66 | |||
XM_021062459.1 | XP_020918118.1 | 574 | 66 | |||
XM_021062460.1 | XP_020918119.1 | 1016 | 66 | |||
XM_021062461.1 | XP_020918120.1 | 556 | 66 | |||
XM_021062462.1 | XP_020918121.1 | 456 | 66 | |||
XM_021062463.1 | XP_020918122.1 | 1067 | 66 | |||
XM_021062464.1 | XP_020918123.1 | 354 | 66 | |||
XM_021062465.1 | XP_020918124.1 | 336 | 66 | |||
XM_021062466.1 | XP_020918125.1 | 1153 | 66 | |||
XM_021062467.1 | XP_020918126.1 | 486 | 66 | |||
XM_021062468.1 | XP_020918127.1 | 233 | 66 | |||
XM_021062469.1 | XP_020918128.1 | 345 | 66 | |||
GenBank mRNA | AK391867.1 | - | 2 - 3 | 172 | 66 | |
AK399686.1 | - | 3 - 4 | 238 | 66 | ||
AY609953.1 | - | 1 - 2 | 258 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
