Assay Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071112.1 | XP_020926771.1 | 271 | 70 | ||
| XM_021071119.1 | XP_020926778.1 | 274 | 70 | |||
| XM_021071124.1 | XP_020926783.1 | 516 | 70 | |||
| GenBank mRNA | AK391958.1 | - | 3 - 4 | 269 | 70 | |
More Information
Set Membership: |
|

SURF1
Genomic Map