Assay Details
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481777.4 | XP_003481825.1 | 534 | 67 | ||
| GenBank mRNA | AK392157.1 | - | 1 - 2 | 491 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EMC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481777.4 | XP_003481825.1 | 534 | 67 | ||
| GenBank mRNA | AK392157.1 | - | 1 - 2 | 491 | 67 | |
Set Membership: |
|