Assay Details
Gene Symbol: RBM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021099250.1 | XP_020954909.1 | 1323 | 58 | ||
XM_021099251.1 | XP_020954910.1 | 1326 | 58 | |||
XM_021099252.1 | XP_020954911.1 | 1279 | 58 | |||
XM_021099253.1 | XP_020954912.1 | 1263 | 58 | |||
XM_021099254.1 | XP_020954913.1 | 4342 | 58 | |||
XM_021099255.1 | XP_020954914.1 | 2968 | 58 | |||
XM_021099256.1 | XP_020954915.1 | 3901 | 58 | |||
XM_021099257.1 | XP_020954916.1 | 2320 | 58 | |||
XM_021099258.1 | XP_020954917.1 | 2268 | 58 | |||
XM_021099259.1 | XP_020954918.1 | 1832 | 58 | |||
XM_021099261.1 | XP_020954920.1 | 1209 | 58 | |||
XM_021099262.1 | XP_020954921.1 | 1150 | 58 | |||
XM_021099263.1 | XP_020954922.1 | 1066 | 58 | |||
XM_021099264.1 | XP_020954923.1 | 1109 | 58 | |||
GenBank mRNA | AK392168.1 | - | 10 - 11 | 1109 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
