Assay Details
| Gene Symbol: DPAGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129927.4 | XP_003129975.1 | 1394 | 64 | ||
| GenBank mRNA | AK392247.1 | - | 8 - 9 | 1319 | 64 | |
More Information
Set Membership: |
|

DPAGT1
Genomic Map