Assay Details
| Gene Symbol: LDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093784.1 | XP_020949443.1 | 1378 | 63 | ||
| GenBank mRNA | AK392264.1 | - | 10 - 11 | 1331 | 63 | |
More Information
Set Membership: |
|

LDHD
Genomic Map