Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003124741.5 | XP_003124789.2 | 926 | 59 | ||
XM_005655185.3 | XP_005655242.1 | 926 | 59 | |||
XM_005655186.3 | XP_005655243.1 | 926 | 59 | |||
XM_013987845.2 | XP_013843299.1 | 872 | 59 | |||
XM_021086763.1 | XP_020942422.1 | 872 | 59 | |||
GenBank mRNA | AK392324.1 | - | 7 - 8 | 899 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
