Assay Details
Gene Symbol: MICOS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005656034.2 | XP_005656091.2 | 223 | 82 | ||
GenBank mRNA | AK392449.1 | - | 1 - 2 | 122 | 82 | |
AY610238.1 | - | 1 - 2 | 109 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
