Assay Details
Gene Symbol: MICOS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005656034.2 | XP_005656091.2 | 274 | 72 | ||
GenBank mRNA | AK392449.1 | - | 2 - 3 | 173 | 72 | |
AY610238.1 | - | 2 - 3 | 160 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
