Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062533.1 | XP_020918192.1 | 606 | 119 | ||
GenBank mRNA | AK392491.1 | - | 3 - 4 | 435 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
