Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021062533.1 | XP_020918192.1 | 652 | 92 | ||
GenBank mRNA | AK392491.1 | - | 4 - 5 | 481 | 92 | |
AK398501.1 | - | 2 - 3 | 122 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
