Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672780.3 | XP_005672837.1 | 1312 | 67 | ||
| XM_005672782.3 | XP_005672839.1 | 1313 | 67 | |||
| XM_021078357.1 | XP_020934016.1 | 1313 | 67 | |||
| GenBank mRNA | AK392511.1 | - | 1 - 2 | 121 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005672780.3, XM_005672782.3, XM_021078357.1 transcripts.
Set Membership: |
|

LZTS3
Genomic Map