Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095660.1 | XP_020951319.1 | 156 | 97 | ||
GenBank mRNA | AK392512.1 | - | 1 - 2 | 79 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021095660.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
