Assay Details
| Gene Symbol: ACHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354481.4 | XP_003354529.1 | 1203 | 94 | ||
| XM_013995487.2 | XP_013850941.2 | 1708 | 94 | |||
| XM_013995488.2 | XP_013850942.2 | 1708 | 94 | |||
| XM_021086251.1 | XP_020941910.1 | 1177 | 94 | |||
| GenBank mRNA | AK392798.1 | - | 2 - 3 | 1181 | 94 | |
More Information
Set Membership: |
|

ACHE
Genomic Map