Assay Details
Gene Symbol: DDX59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005653796.3 | XP_005653853.1 | 1823 | 72 | ||
XM_005653797.3 | XP_005653854.1 | 1751 | 72 | |||
XM_005653799.3 | XP_005653856.1 | 1656 | 72 | |||
XM_013978422.2 | XP_013833876.1 | 1825 | 72 | |||
XM_021064498.1 | XP_020920157.1 | 2333 | 72 | |||
XM_021064499.1 | XP_020920158.1 | 1576 | 72 | |||
GenBank mRNA | AK392886.1 | - | 4 - 5 | 961 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
