Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021096606.1 | XP_020952265.1 | 1208 | 68 | ||
XM_021096607.1 | XP_020952266.1 | 906 | 68 | |||
GenBank mRNA | AK393080.1 | - | 3 - 4 | 243 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
