Assay Details
Gene Symbol: LRRC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021069763.1 | XP_020925422.1 | 371 | 97 | ||
XM_021069764.1 | XP_020925423.1 | 365 | 97 | |||
XM_021069765.1 | XP_020925424.1 | 371 | 97 | |||
XM_021069766.1 | XP_020925425.1 | 371 | 97 | |||
XM_021069767.1 | XP_020925426.1 | 220 | 97 | |||
GenBank mRNA | AK393341.1 | - | 3 - 4 | 367 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
