Assay Details
| Gene Symbol: METTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096140.1 | XP_020951799.1 | 1965 | 73 | ||
| XM_021096141.1 | XP_020951800.1 | 1980 | 73 | |||
| XM_021096146.1 | XP_020951805.1 | 1691 | 73 | |||
| XM_021096147.1 | XP_020951806.1 | 1705 | 73 | |||
| GenBank mRNA | AK394672.1 | - | 1 | 317 | 73 | |
More Information
Set Membership: |
|

METTL4
Genomic Map