Assay Details
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013994460.2 | XP_013849914.1 | 80 | |||
XM_021083210.1 | XP_020938869.1 | 80 | ||||
XM_021083211.1 | XP_020938870.1 | 80 | ||||
XM_021083212.1 | XP_020938871.1 | 80 | ||||
XM_021083213.1 | XP_020938872.1 | 80 | ||||
XM_021083215.1 | XP_020938874.1 | 80 | ||||
GenBank mRNA | AK394704.1 | - | 5 - 6 | 855 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
