Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925977.5 | XP_001926012.1 | 1058 | 71 | ||
| XM_005670673.3 | XP_005670730.1 | 1196 | 71 | |||
| XM_005670674.3 | XP_005670731.1 | 551 | 71 | |||
| XM_021073844.1 | XP_020929503.1 | 1132 | 71 | |||
| GenBank mRNA | AK394713.1 | - | 5 - 6 | 587 | 71 | |
More Information
Set Membership: |
|

WSB2
Genomic Map