Assay Details
| Gene Symbol: BCL7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483424.4 | XP_003483472.1 | 3273 | 76 | ||
| XM_021074246.1 | XP_020929905.1 | 3985 | 76 | |||
| GenBank mRNA | AK394920.1 | - | 1 | 823 | 76 | |
More Information
Set Membership: |
|

BCL7A
Genomic Map