Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124608.6 | XP_003124656.3 | 391 | 93 | ||
| XM_005662147.3 | XP_005662204.1 | 410 | 93 | |||
| XM_013995659.2 | XP_013851113.2 | 387 | 93 | |||
| GenBank mRNA | AK394945.1 | - | 3 - 4 | 329 | 93 | |
| AY609426.1 | - | 3 - 4 | 318 | 93 | ||
More Information
Set Membership: |
|

CARHSP1
Genomic Map