Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655394.3 | XP_005655451.1 | 515 | 71 | ||
| XM_005655395.3 | XP_005655452.1 | 703 | 71 | |||
| XM_021089912.1 | XP_020945571.1 | 2842 | 71 | |||
| XM_021089913.1 | XP_020945572.1 | 2957 | 71 | |||
| GenBank mRNA | AK394979.1 | - | 2 - 3 | 681 | 71 | |
More Information
Set Membership: |
|

CIART
Genomic Map