Assay Details
| Gene Symbol: NSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100910.1 | XP_020956569.1 | 4086 | 93 | ||
| XM_021100911.1 | XP_020956570.1 | 4163 | 93 | |||
| XM_021100913.1 | XP_020956572.1 | 3894 | 93 | |||
| XM_021100914.1 | XP_020956573.1 | 3209 | 93 | |||
| GenBank mRNA | AK395014.1 | - | 2 - 3 | 294 | 93 | |
More Information
Set Membership: |
|

NSD2
Genomic Map