Assay Details
| Gene Symbol: PMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013984337.2 | XP_013839791.2 | 2093 | 84 | ||
| XM_021075389.1 | XP_020931048.1 | 2045 | 84 | |||
| XM_021075390.1 | XP_020931049.1 | 2100 | 84 | |||
| XM_021075391.1 | XP_020931050.1 | 2043 | 84 | |||
| XM_021075392.1 | XP_020931051.1 | 2035 | 84 | |||
| XM_021075393.1 | XP_020931052.1 | 2009 | 84 | |||
| XM_021075394.1 | XP_020931053.1 | 2043 | 84 | |||
| XM_021075395.1 | XP_020931054.1 | 2163 | 84 | |||
| XM_021075396.1 | XP_020931055.1 | 1990 | 84 | |||
| XM_021075397.1 | XP_020931056.1 | 1875 | 84 | |||
| XM_021075398.1 | XP_020931057.1 | 1507 | 84 | |||
| XM_021075399.1 | XP_020931058.1 | 2037 | 84 | |||
| XM_021075400.1 | XP_020931059.1 | 1606 | 84 | |||
| XM_021075401.1 | XP_020931060.1 | 1428 | 84 | |||
| GenBank mRNA | AK395067.1 | - | 10 - 11 | 2037 | 84 | |
More Information
Set Membership: |
|

PMS1
Genomic Map