Assay Details
| Gene Symbol: GIPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083694.1 | XP_020939353.1 | 1043 | 84 | ||
| XM_021083695.1 | XP_020939354.1 | 1072 | 84 | |||
| XM_021083696.1 | XP_020939355.1 | 1058 | 84 | |||
| GenBank mRNA | AK395337.1 | - | 8 - 9 | 981 | 84 | |
More Information
Set Membership: |
|

GIPC1
Genomic Map