Assay Details
| Gene Symbol: PRDM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083147.1 | XP_020938806.1 | 694 | 71 | ||
| XM_021083148.1 | XP_020938807.1 | 468 | 71 | |||
| XM_021083149.1 | XP_020938808.1 | 710 | 71 | |||
| XM_021083150.1 | XP_020938809.1 | 258 | 71 | |||
| XM_021083151.1 | XP_020938810.1 | 694 | 71 | |||
| XM_021083152.1 | XP_020938811.1 | 592 | 71 | |||
| XM_021083153.1 | XP_020938812.1 | 468 | 71 | |||
| GenBank mRNA | AK395430.1 | - | 3 - 4 | 468 | 71 | |
More Information
Set Membership: |
|

PRDM11
Genomic Map