Assay Details
| Gene Symbol: SLC7A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094151.1 | XP_020949810.1 | 966 | 102 | ||
| XM_021094152.1 | XP_020949811.1 | 868 | 102 | |||
| XM_021094153.1 | XP_020949812.1 | 760 | 102 | |||
| XM_021094154.1 | XP_020949813.1 | 862 | 102 | |||
| XM_021094155.1 | XP_020949814.1 | 831 | 102 | |||
| XM_021094156.1 | XP_020949815.1 | 1778 | 102 | |||
| XM_021094157.1 | XP_020949816.1 | 1493 | 102 | |||
| GenBank mRNA | AK395847.1 | - | 2 - 3 | 585 | 102 | |
More Information
Set Membership: |
|

SLC7A6
Genomic Map