Assay Details
Gene Symbol: DAXX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005665885.3 | XP_005665942.1 | 2179 | 94 | ||
XM_005665886.3 | XP_005665943.1 | 2149 | 94 | |||
XM_013977770.2 | XP_013833224.1 | 2480 | 94 | |||
XM_013977771.2 | XP_013833225.1 | 2071 | 94 | |||
XM_021098591.1 | XP_020954250.1 | 2131 | 94 | |||
GenBank mRNA | AK396226.1 | - | 4 - 5 | 1212 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
